3-Hydroxy-3-phenylpropansäure 98%

Lieferant: Apollo Scientific

Synonyme: 3-Hydroxy-3-phenylpropionic acid

OR902414-25G
APOSOR902414-25GEA 116 EUR
APOSOR902414-25G
3-Hydroxy-3-phenylpropansäure 98%
3-Hydroxy-3-phenylpropanoic acid
Formel: C9H10O3
Molecular Weight: 166.1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00137907
CAS-Nummer: 3480-87-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR