5-Aminopyridine-2-thiol 95%

Lieferant: Apollo Scientific
Achtung

OR902423-250MG
APOSOR902423-250MGEA 319 EUR
APOSOR902423-250MG
5-Aminopyridine-2-thiol 95%
5-Aminopyridine-2-thiol
Formel: C₅H₆N₂S
Molecular Weight: 126,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09029363
CAS-Nummer: 27885-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR