N1,N1,N2-Trimethylethane-1,2-diamine dihydrochloride 96%

Lieferant: Apollo Scientific

OR902448-5G
APOSOR902448-5GEA 87 EUR
APOSOR902448-5G
N1,N1,N2-Trimethylethane-1,2-diamine dihydrochloride 96%
N1,N1,N2-Trimethylethane-1,2-diamine dihydrochloride
Formel: C₅H₁₆Cl₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD21604009
CAS-Nummer: 326888-32-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR