2-Methoxy-5-methylpyridin-3-amine 98%
Lieferant: Apollo Scientific
OR902464-1G
APOSOR902464-1GEA
632
EUR
APOSOR902464-1G
2-Methoxy-5-methylpyridin-3-amine 98%
2-Methoxy-5-methylpyridin-3-amine
Formel:
C₇H₁₀N₂O Molecular Weight: 138,167 g/mol Schmelzpunkt: 58…59 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD13185338 CAS-Nummer: 179677-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...