5-Chloro-2-ethoxypyridin-3-amine 95%

Lieferant: Apollo Scientific

OR902505-1G
APOSOR902505-1GEA 632 EUR
APOSOR902505-1G
5-Chloro-2-ethoxypyridin-3-amine 95%
5-Chloro-2-ethoxypyridin-3-amine
Formel: C₇H₉ClN₂O
Molecular Weight: 172,612 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07375047
CAS-Nummer: 886373-74-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR