2-(3,5-Dimethylphenyl)piperidine 95%
Lieferant: Apollo Scientific
OR902522-1G
APOSOR902522-1GEA
74.1
EUR
APOSOR902522-1G
2-(3,5-Dimethylphenyl)piperidine 95%
2-(3,5-Dimethylphenyl)piperidine
Formel:
C₁₃H₁₉N Molecular Weight: 189,297 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD02663617 CAS-Nummer: 383128-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...