1-(2-Methylpyridin-4-yl)ethanone 95%

Lieferant: Apollo Scientific

Synonyme: 4-Acetyl-2-methylpyridine

OR902630-1G
APOSOR902630-1GEA 108 EUR
APOSOR902630-1G
1-(2-Methylpyridin-4-yl)ethanone 95%
1-(2-Methylpyridin-4-yl)ethanone
Formel: C₈H₉NO
Molecular Weight: 135,1632 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07375079
CAS-Nummer: 2732-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR