N-(4-Hydroxyphenyl)maleimide 98%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Hydroxyphenyl)-1H-pyrrole-2,5-dione

OR902636-100G
APOSOR902636-100GEA 148 EUR
APOSOR902636-100G
N-(4-Hydroxyphenyl)maleimide 98%
N-(4-Hydroxyphenyl)maleimide
Formel: C₁₀H₇NO₃
Molecular Weight: 189.167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00129167
CAS-Nummer: 7300-91-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR