N-Isopropylanilin 98%

Lieferant: Apollo Scientific

OR902721-100G
APOSOR902721-100GEA 46.4 EUR
APOSOR902721-100G
N-Isopropylanilin 98%
N-Isopropylanilin
Formel: C₆H₅NHCH(CH₃)₂
Molecular Weight: 135,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00026347
CAS-Nummer: 768-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR