3'-(Methylthio)acetanilid 95%
Lieferant: Apollo Scientific
OR902818-25G
OR902818-5G
APOSOR902818-25GEA
400
EUR
APOSOR902818-25G
APOSOR902818-5G
3'-(Methylthio)acetanilid 95%
3'-(Methylthio)acetanilid
Formel:
C₉H₁₁NOS Lagertemperatur: Raumtemperatur |
CAS-Nummer:
2524-78-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...