3'-(Methylthio)acetanilid 95%

Lieferant: Apollo Scientific

OR902818-25G OR902818-5G
APOSOR902818-25GEA 400 EUR
APOSOR902818-25G APOSOR902818-5G
3'-(Methylthio)acetanilid 95%
3'-(Methylthio)acetanilid
Formel: C₉H₁₁NOS
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2524-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR