tert-Butyl (2-oxopropyl)carbamate 97%

Lieferant: Apollo Scientific

Synonyme: (2-Oxopropyl)carbamic acid tert-butyl ester

OR902921-25G
APOSOR902921-25GEA 84.1 EUR
APOSOR902921-25G
tert-Butyl (2-oxopropyl)carbamate 97%
tert-Butyl (2-oxopropyl)carbamate
Formel: C₈H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD10698678
CAS-Nummer: 170384-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR