5-(Aminomethyl)-2-methyl-4-pyrimidinamin 96%
Lieferant: Apollo Scientific
Synonyme:
Grewe diamine, 4-Amino-5-aminomethyl-2-methylpyrimidine
OR902949-5G
APOSOR902949-5GEA
198
EUR
APOSOR902949-5G
5-(Aminomethyl)-2-methyl-4-pyrimidinamin 96%
5-(Aminomethyl)-2-methylpyrimidin-4-amine
Formel:
C₆H₁₀N₄ Schmelzpunkt: 270…272 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00078880 CAS-Nummer: 95-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...