2-Methyl-6-(2-pyrrolidinylmethyl)pyridine 95%

Lieferant: Apollo Scientific

OR902951-100MG
APOSOR902951-100MGEA 957 EUR
APOSOR902951-100MG
2-Methyl-6-(2-pyrrolidinylmethyl)pyridine 95%
2-Methyl-6-(2-pyrrolidinylmethyl)pyridine
Formel: C₁₁H₁₆N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05189307
CAS-Nummer: 524674-47-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR