2-(4-Methylphenyl)pyrrolidine 98%

Lieferant: Apollo Scientific

OR902962-10G OR902962-1G OR902962-5G
APOSOR902962-10GEA 328 EUR
APOSOR902962-10G APOSOR902962-1G APOSOR902962-5G
2-(4-Methylphenyl)pyrrolidine 98%
2-(4-Methylphenyl)pyrrolidine
Formel: C₁₁H₁₅N
Lagertemperatur: Raumtemperatur
MDL: MFCD02663452
CAS-Nummer: 62506-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR