2-(tert-Butoxy)aniline 95%

Lieferant: Apollo Scientific

OR902981-1G
APOSOR902981-1GEA 451 EUR
APOSOR902981-1G
2-(tert-Butoxy)aniline 95%
2-(tert-Butoxy)aniline
Formel: C₁₀H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD11196192
CAS-Nummer: 127517-27-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR