(3,5-Dimethylpyrazol-1-yl)acetic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-(3,5-Dimethyl-1H-pyrazol-1-yl)acetic acid

OR903023-25G
APOSOR903023-25GEA 264 EUR
APOSOR903023-25G
(3,5-Dimethylpyrazol-1-yl)acetic acid 97%
(3,5-Dimethylpyrazol-1-yl)acetic acid
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00297249
CAS-Nummer: 16034-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR