2,2-Dimethoxyethyl isocyanide 95%

Lieferant: Apollo Scientific

OR903045-1G
APOSOR903045-1GEA 538 EUR
APOSOR903045-1G
2,2-Dimethoxyethyl isocyanide 95%
2,2-Dimethoxyethyl isocyanide
Formel: C₅H₉NO₂
Molecular Weight: 115,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664543
CAS-Nummer: 277300-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR