2-Amino-4-ethyl-5-methylthiophene-3-carbonitrile 95%

Lieferant: Apollo Scientific

OR903049-1G OR903049-250MG
APOSOR903049-1GEA 683 EUR
APOSOR903049-1G APOSOR903049-250MG
2-Amino-4-ethyl-5-methylthiophene-3-carbonitrile 95%
2-Amino-4-ethyl-5-methylthiophene-3-carbonitrile
Formel: C₈H₁₀N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD01114970
CAS-Nummer: 4651-92-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR