2,4,6-Trimethyl-m-phenylendiamin

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4,6-Trimethylbenzol-1,3-diamin, 2,4-Diamino-1,3,5-trimethylbenzol, 1,3-Diamino-2,4,6-trimethylbenzol, 2,4,6-Trimethyl-1,3-phenylendiamin

OR903076-25G OR903076-100G
APOSOR903076-25GEA 65.4 EUR
APOSOR903076-25G APOSOR903076-100G
2,4,6-Trimethyl-m-phenylendiamin
2,4,6-Trimethyl-m-phenylendiamin
Formel: C₉H₁₄N₂
Molecular Weight: 150.22 g/mol
Schmelzpunkt: 89…91 °C
MDL: MFCD00010149
CAS-Nummer: 3102-70-3
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR