2,4,6-Trimethyl-m-phenylendiamin

Lieferant: Apollo Scientific
Achtung

OR903076-25G OR903076-100G
APOSOR903076-25GEA 65.4 EUR
APOSOR903076-25G APOSOR903076-100G
2,4,6-Trimethyl-m-phenylendiamin
2,4,6-Trimethyl-m-phenylendiamin
Formel: C₉H₁₄N₂
Schmelzpunkt: 89 - 91 °C
MDL: MFCD00010149
CAS-Nummer: 3102-70-3
UN: 3077
ADR: 9+EHS,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR