3,4-Xylylessigsäure 95%
Lieferant: Apollo Scientific
Synonyme:
3,4-Dimethylphenylacetic acid
OR903130-5G
APOSOR903130-5GEA
59.6
EUR
APOSOR903130-5G
3,4-Xylylessigsäure 95%
3,4-Xylylessigsäure
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164.2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02664684 CAS-Nummer: 17283-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...