Ethyl-5-chlor-2-thiophencarboxylat 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 5-chlorothiophene-2-carboxylate

OR903157-25G
APOSOR903157-25GEA 120 EUR
APOSOR903157-25G
Ethyl-5-chlor-2-thiophencarboxylat 97%
Ethyl-5-chlor-2-thiophencarboxylat
Formel: C₇H₇ClO₂S
Molecular Weight: 190.65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051732
CAS-Nummer: 5751-82-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR