2-(4-Methoxyphenyl)-2-methylpropylamine 98%

Lieferant: Apollo Scientific

OR903208-1G
APOSOR903208-1GEA 299 EUR
APOSOR903208-1G
2-(4-Methoxyphenyl)-2-methylpropylamine 98%
2-(4-Methoxyphenyl)-2-methylpropylamine
Formel: C₁₁H₁₇NO
Molecular Weight: 179,259 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01937048
CAS-Nummer: 51558-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR