2-Methyl-4-(4-methylphenyl)-1,3-thiazole 95%

Lieferant: Apollo Scientific
Achtung

OR903268-1G
APOSOR903268-1GEA 219 EUR
APOSOR903268-1G
2-Methyl-4-(4-methylphenyl)-1,3-thiazole 95%
2-Methyl-4-(4-methylphenyl)-1,3-thiazole
Formel: C₁₁H₁₁NS
Molecular Weight: 189,277 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00662551
CAS-Nummer: 66047-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR