2,3-Dihydro-1-oxo-1H-indene-5-carbonitrile 95%

Lieferant: Apollo Scientific

OR903289-1G
APOSOR903289-1GEA 74.1 EUR
APOSOR903289-1G
2,3-Dihydro-1-oxo-1H-indene-5-carbonitrile 95%
2,3-Dihydro-1-oxo-1H-indene-5-carbonitrile
Formel: C₁₀H₇NO
Molecular Weight: 157.1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08234685
CAS-Nummer: 25724-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR