1-(Methoxymethyl)-3-methyl-2-nitrobenzene 96%
Lieferant: Apollo Scientific
OR903303-1G
APOSOR903303-1GEA
502
EUR
APOSOR903303-1G
1-(Methoxymethyl)-3-methyl-2-nitrobenzene 96%
1-(Methoxymethyl)-3-methyl-2-nitrobenzene
Formel:
C₉H₁₁NO₃ Molecular Weight: 181,189 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22420432 CAS-Nummer: 507455-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...