5-Chinoxalinol 98%

Lieferant: Apollo Scientific

OR903311-5G
APOSOR903311-5GEA 163 EUR
APOSOR903311-5G
5-Chinoxalinol 98%
Quinoxalin-5-ol
Formel: C8H6N2O
Lagertemperatur: Raumtemperatur
MDL: MFCD00972004
CAS-Nummer: 17056-99-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR