Ethyl-6-chloro-3-pyridazinecarboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl 6-chloro-3-pyridazinecarboxylate, Ethyl 6-chloropyridazine-3-carboxylate, 6-chloro-3-pyridazinecarboxylic acid ethyl ester, 6-Chloro-pyridazine-3-carboxylic acid ethyl ester

OR903332-5G
APOSOR903332-5GEA 139 EUR
APOSOR903332-5G
Ethyl-6-chloro-3-pyridazinecarboxylate 98%
Ethyl-6-chloro-3-pyridazinecarboxylate
Formel: C₇H₇ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09908168
CAS-Nummer: 75680-92-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR