tert-Butyl N-(1-cyano-1-methylethyl)carbamate 95%

Lieferant: Apollo Scientific

OR903352-1G
APOSOR903352-1GEA 538 EUR
APOSOR903352-1G
tert-Butyl N-(1-cyano-1-methylethyl)carbamate 95%
tert-Butyl N-(1-cyano-1-methylethyl)carbamate
Formel: C₉H₁₆N₂O₂
Molecular Weight: 184,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12791990
CAS-Nummer: 133117-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR