N-(2-Cyanophenyl)acetamide 97%

Lieferant: Apollo Scientific

OR903353-1G OR903353-250MG
APOSOR903353-1GEA 596 EUR
APOSOR903353-1G APOSOR903353-250MG
N-(2-Cyanophenyl)acetamide 97%
N-(2-Cyanophenyl)acetamide
Formel: C₉H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00222087
CAS-Nummer: 25116-00-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR