6-Chloropyrido[3,4-d]pyrimidin-4(3H)-one 98%

Lieferant: Apollo Scientific

OR903355-1G
APOSOR903355-1GEA 62.5 EUR
APOSOR903355-1G
6-Chloropyrido[3,4-d]pyrimidin-4(3H)-one 98%
6-Chloropyrido[3,4-d]pyrimidin-4(3H)-one
Formel: C₇H₄ClN₃O
Molecular Weight: 181.5800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044635
CAS-Nummer: 171178-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR