Thioisonicotinamid 98%

Lieferant: Apollo Scientific

OR903374-100G
APOSOR903374-100GEA 84.1 EUR
APOSOR903374-100G
Thioisonicotinamid 98%
Thioisonicotinamid
Formel: C₆H₆N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00006437
CAS-Nummer: 2196-13-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR