(4-Methoxybutyl)boronic acid 95%
Lieferant: Apollo Scientific
OR903421-1G
APOSOR903421-1GEA
632
EUR
APOSOR903421-1G
(4-Methoxybutyl)boronic acid 95%
(4-Methoxybutyl)boronic acid
Formel:
C₅H₁₃BO₃ Molecular Weight: 131.966 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26926052 CAS-Nummer: 1919893-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...