2-Benzotriazol-1-yl-acetamide 95%
Lieferant: Apollo Scientific
OR903504-1G
APOSOR903504-1GEA
647
EUR
APOSOR903504-1G
2-Benzotriazol-1-yl-acetamide 95%
2-Benzotriazol-1-yl-acetamide
Formel:
C₈H₈N₄O Molecular Weight: 176,175 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00964268 CAS-Nummer: 69218-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...