4-Cyano-2,6-dimethylphenylboronic acid 95%
Lieferant: Apollo Scientific
OR903551-1G
APOSOR903551-1GEA
806
EUR
APOSOR903551-1G
4-Cyano-2,6-dimethylphenylboronic acid 95%
4-Cyano-2,6-dimethylphenylboronic acid
Formel:
C₉H₁₀BNO₂ Molecular Weight: 174,992 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22056751 CAS-Nummer: 1451391-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...