Ethyl-2-(methylamino)benzoate 95%
Lieferant: Apollo Scientific
OR903561-1G
APOSOR903561-1GEA
150
EUR
APOSOR903561-1G
Ethyl-2-(methylamino)benzoate 95%
Ethyl-2-(methylamino)benzoate
Formel:
C₁₀H₁₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11174593 CAS-Nummer: 35472-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...