3,4-Dichlor-1,2,5-thiadiazol 98%

Lieferant: Apollo Scientific

OR903574-100G
APOSOR903574-100GEA 75.4 EUR
APOSOR903574-100G
3,4-Dichlor-1,2,5-thiadiazol 98%
3,4-Dichloro-1,2,5-thiadiazole
Formel: C₂Cl₂N₂S
Molecular Weight: 155.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010072
CAS-Nummer: 5728-20-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR