2-Chloro-N-ethylacetamide 97%

Lieferant: Apollo Scientific

OR903605-5G
APOSOR903605-5GEA 125 EUR
APOSOR903605-5G
2-Chloro-N-ethylacetamide 97%
2-Chloro-N-ethylacetamide
Formel: C₄H₈ClNO
Molecular Weight: 121,565 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040868
CAS-Nummer: 105-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR