1-Isocyano-3,5-dimethyl-benzene 95%
Lieferant: Apollo Scientific
OR903616-1G
APOSOR903616-1GEA
348
EUR
APOSOR903616-1G
1-Isocyano-3,5-dimethyl-benzene 95%
1-Isocyano-3,5-dimethyl-benzene
Formel:
C₉H₉N Molecular Weight: 131,174 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02664617 CAS-Nummer: 20600-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...