5-Nitro-m-toluidin 96%

Lieferant: Apollo Scientific

Synonyme: 3-Methyl-5-nitroaniline

OR903637-5G
APOSOR903637-5GEA 143 EUR
APOSOR903637-5G
5-Nitro-m-toluidin 96%
5-Nitro-m-toluidin
Formel: C₇H₈N₂O₂
Molecular Weight: 152.1500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082655
CAS-Nummer: 618-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR