5-Aminoimidazo[1,2-a]pyridin 95%

Lieferant: Apollo Scientific

OR903639-5G
APOSOR903639-5GEA 68.3 EUR
APOSOR903639-5G
5-Aminoimidazo[1,2-a]pyridin 95%
5-Aminoimidazo[1,2-a]pyridin
Formel: C₇H₇N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05863377
CAS-Nummer: 66358-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR