2-Chloro-4-ethynylpyridine 98%

Lieferant: Apollo Scientific

OR903659-5G
APOSOR903659-5GEA 255 EUR
APOSOR903659-5G
2-Chloro-4-ethynylpyridine 98%
2-Chloro-4-ethynylpyridine
Formel: C₇H₄ClN
Molecular Weight: 137.5664 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189742
CAS-Nummer: 945717-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR