1-Methyl-1,2,3,6-tetrahydropyridine hydrochloride 95%

Lieferant: Apollo Scientific

OR903703-1G
APOSOR903703-1GEA 0 EUR
APOSOR903703-1G
1-Methyl-1,2,3,6-tetrahydropyridine hydrochloride 95%
1-Methyl-1,2,3,6-tetrahydropyridine hydrochloride
Formel: C₆H₁₂ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD00074973
CAS-Nummer: 73107-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR