5-Chlorothiophene-3-carboxylic acid 96%

Lieferant: Apollo Scientific

OR903783-5G
APOSOR903783-5GEA 163 EUR
APOSOR903783-5G
5-Chlorothiophene-3-carboxylic acid 96%
5-Chlorothiophene-3-carboxylic acid
Formel: C5H3ClO2S
Molecular Weight: 162.5900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082877
CAS-Nummer: 36157-42-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR