4H-Cyclopenta[b]thiophen-6(5H)-one 98%

Lieferant: Apollo Scientific

OR903852-1G
APOSOR903852-1GEA 116 EUR
APOSOR903852-1G
4H-Cyclopenta[b]thiophen-6(5H)-one 98%
4H-Cyclopenta[b]thiophen-6(5H)-one
Formel: C₇H₆OS
Molecular Weight: 138,1869 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03426936
CAS-Nummer: 5650-52-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR