3-Methoxy-4-nitrophenol 96%

Lieferant: Apollo Scientific

OR903872-5G
APOSOR903872-5GEA 108 EUR
APOSOR903872-5G
3-Methoxy-4-nitrophenol 96%
3-Methoxy-4-nitrophenol
Formel: C7H7NO4
Lagertemperatur: Raumtemperatur
MDL: MFCD11840329
CAS-Nummer: 16292-95-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR