Methyl-2-aminobenzo[d]thiazole-7-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

OR903880-1G OR903880-5G
APOSOR903880-1GEA 0 EUR
APOSOR903880-1G APOSOR903880-5G
Methyl-2-aminobenzo[d]thiazole-7-carboxylate 98%
Methyl-2-aminobenzo[d]thiazole-7-carboxylate
Formel: C₉H₈N₂O₂S
Lagertemperatur: Kühlschrank
MDL: MFCD09263762
CAS-Nummer: 209459-11-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR