5-Brom-2-pentanon 95%

Lieferant: Apollo Scientific

OR903890-10G OR903890-1G OR903890-5G
APOSOR903890-10GEA 632 EUR
APOSOR903890-10G APOSOR903890-1G APOSOR903890-5G
5-Brom-2-pentanon 95%
5-Brom-2-pentanon
Formel: H₃CC(O)(CH₂)₂CH₂Br
Lagertemperatur: Raumtemperatur
MDL: MFCD00051425
CAS-Nummer: 3884-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR