2-Morpholinopyrimidin-4-ol 95%

Lieferant: Apollo Scientific

OR903939-1G
APOSOR903939-1GEA 71.2 EUR
APOSOR903939-1G
2-Morpholinopyrimidin-4-ol 95%
2-Morpholinopyrimidin-4-ol
Formel: C₈H₁₁N₃O₂
Molecular Weight: 181,1900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234443
CAS-Nummer: 19810-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR